missing translation for 'onlineSavingsMsg'
Learn More

Potassium Benzoate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18612458
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612458 12 kg Poly Bottle
18612456 500 g Poly Bottle
18612457 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612458 Supplier Spectrum Chemical Mfg Cor Supplier No. P117312KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 582-25-2

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 12 kg
Molecular Weight (g/mol) 160.22
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Benzoate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.