accessibility menu, dialog, popup

UserName

CAS RN 58109-40-3

IUPAC Name: diphenyliodanium
Synonyms: diphenyliodanium
Molecular Weight (g/mol): 281.12
Molecular Formula: C12H10I
InChi Key: OZLBDYMWFAHSOQ-UHFFFAOYSA-N
SMILES: [I+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)