accessibility menu, dialog, popup

UserName

CAS RN 58-33-3

IUPAC Name: hydrogen dimethyl[1-(10H-phenothiazin-10-yl)propan-2-yl]amine chloride
Synonyms: hydrogen promethazine chloride
Molecular Weight (g/mol): 320.88
Molecular Formula: C17H21ClN2S
InChi Key: XXPDBLUZJRXNNZ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(CN1C2=CC=CC=C2SC2=CC=CC=C12)N(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)