missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride 98.0+%, TCI America™

Catalog No. P202925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P202925G 25 g
P2029100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P202925G Supplier TCI America Supplier No. P202925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-33-3
Molecular Formula C17H21ClN2S
Molecular Weight (g/mol) 320.879
MDL Number MFCD00012652
InChI Key XXPDBLUZJRXNNZ-UHFFFAOYSA-N
Synonym promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen
PubChem CID 6014
ChEBI CHEBI:8462
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
SMILES CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 320.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Promethazine Hydrochloride
RTECSNumber SO8225000
TSCA Yes