missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Promethazine hydrochloride

Catalog No. AC254581000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC254581000 100 g
AC254580050 5 g
AC254580250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC254581000 Supplier Thermo Scientific Chemicals Supplier No. 254581000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-33-3 | C17H20N2S·HCl | 320.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-33-3
Molecular Formula C17H20N2S·HCl
Molecular Weight (g/mol) 320.88
InChI Key XXPDBLUZJRXNNZ-UHFFFAOYSA-N
Synonym promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen
PubChem CID 6014
ChEBI CHEBI:8462
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
SMILES CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl

Specifications

Melting Point 227.0°C to 230.0°C
Quantity 100 g
Solubility Information (10% in water) Clear colorless to light yellow
Formula Weight 320.88
Chemical Name or Material Promethazine hydrochloride
Hazard Category GHS Signal Word: Warning
EINECSNumber 200-375-2

RUO – Research Use Only