missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019454
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019454 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019454 Supplier Merck Emd Millipore Supplier No. PHR14671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Promethazine hydrochloride belongs to the group of phenothiazine derivatives. It is commonly used as an antihistaminic for most prominently enhancing the anesthetic, sedative and analgesic effect of other drugs. It also shows antipyretic and antiemetic properties, when used in combination with paracetamol. It is also used to provide symptomatic relief of hypersensitivity reactions.

Specifications

CAS 58-33-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H20N2 S · HCl
MDL Number MFCD00012652
Synonym Promethazine hydrochloride; 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 320.88
Grade Certified Reference Material