accessibility menu, dialog, popup

UserName

CAS RN 5793-85-1

IUPAC Name: calcium benzene-1,2-dicarboxylate
Synonyms: calcium phthalate
Molecular Weight (g/mol): 204.19
Molecular Formula: C8H4CaO4
InChi Key: PYSZASIZWHHPHJ-UHFFFAOYSA-L
SMILES: [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results