missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phthalate Hydrate 98.0+%, TCI America™

Catalog No. C20035G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20035G Supplier TCI America Supplier No. C20035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5793-85-1
Molecular Formula C8H4CaO4
Molecular Weight (g/mol) 204.19
MDL Number MFCD08276328
InChI Key PYSZASIZWHHPHJ-UHFFFAOYSA-L
Synonym Phthalic Acid Calcium Salt
PubChem CID 91659099
IUPAC Name calcium benzene-1,2-dicarboxylate
SMILES [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 204.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Phthalate Hydrate
EINECSNumber (3)-1272
TSCA No