CAS RN 57644-54-9
IUPAC Name:
bismuth(3+) 3,4-dicarboxy-3-hydroxybutanoate 3,4-dicarboxy-3-oxidobutanoate tripotassium
Synonyms:
bismuth(3+) 3,4-dicarboxy-3-hydroxybutanoate 3,4-dicarboxy-3-oxidobutanoate tripotassium
Molecular Weight (g/mol):
707.5
Molecular Formula:
C12H13BiK3O14
InChi Key:
BJTMZHBPYPNQSV-UHFFFAOYNA-L
SMILES:
[K].[K].[K].[Bi+3].OC(=O)CC(O)(CC([O-])=O)C(O)=O.OC(=O)CC([O-])(CC([O-])=O)C(O)=O
1
–
1
of
1
results
1
Bismuth Tripotassium Dicitrate, TCI America™
CAS: 57644-54-9 Molecular Formula: C12H10BiK3O14 Molecular Weight (g/mol): 704.473 MDL Number: MFCD02101411 InChI Key: ZQUAVILLCXTKTF-UHFFFAOYSA-H PubChem CID: 10101269 IUPAC Name: bismuth;tripotassium;2-hydroxypropane-1,2,3-tricarboxylate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[K+].[K+].[K+].[Bi+3]