missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Tripotassium Dicitrate, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 704.47 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bismuth Tripotassium Dicitrate |
Safety and Handling
| RTECSNumber | TZ8492900 |
| TSCA | No |