accessibility menu, dialog, popup

UserName

CAS RN 5763-61-1

IUPAC Name: 1-(3,4-dimethoxyphenyl)methanamine
Synonyms: 3,4-dimethoxybenzylamine
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: DIVNUTGTTIRPQA-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN)C=C1OC
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)