accessibility menu, dialog, popup

UserName

CAS RN 5762-56-1

IUPAC Name: [bis(dimethylazaniumyl)methyl]dimethylazanium
Synonyms: [bis(dimethylammonio)methyl]dimethylazanium
Molecular Weight (g/mol): 148.27
Molecular Formula: C7H22N3
InChi Key: MUMVIYLVHVCYGI-UHFFFAOYSA-Q
SMILES: C[NH+](C)C([NH+](C)C)[NH+](C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results