missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)methane (stabilized with KOH) 97.0+%, TCI America™

Catalog No. T10575ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10575ML 5 mL
T105725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10575ML Supplier TCI America Supplier No. T10575ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5762-56-1
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008322
InChI Key MUMVIYLVHVCYGI-UHFFFAOYSA-N
Synonym tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
PubChem CID 79831
IUPAC Name N,N,N',N',N'',N''-hexamethylmethanetriamine
SMILES CN(C)C(N(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 mL
UN Number 2733
Formula Weight 145.25
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(dimethylamino)methane (stabilized with KOH)