accessibility menu, dialog, popup

UserName

CAS RN 576-83-0

IUPAC Name: 2-bromo-1,3,5-trimethylbenzene
Synonyms: 2-bromo-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: RRTLQRYOJOSPEA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C(C)=C1
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)