missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Bromide 99.0+%, TCI America™

Catalog No. B1261500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1261500G 500 g
B126125G 25 g
B1261100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1261500G Supplier TCI America Supplier No. B1261500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 576-83-0
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00000073
InChI Key RRTLQRYOJOSPEA-UHFFFAOYSA-N
Synonym 2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene
PubChem CID 68473
IUPAC Name 2-bromo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)Br)C

Specifications

Color Yellow
Boiling Point 225°C
Quantity 500 g
Formula Weight 199.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Mesityl Bromide
EINECSNumber (3)-0057
TSCA Yes