accessibility menu, dialog, popup

UserName

H3COPOOOOCH3

CAS RN 57443-18-2

IUPAC Name: benzyl 2-(dimethoxyphosphoryl)acetate
Synonyms: benzyl 2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 258.21
Molecular Formula: C11H15O5P
InChi Key: QYLGNJMIOVHLQQ-UHFFFAOYSA-N
SMILES: COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)