accessibility menu, dialog, popup

UserName

CAS RN 573-57-9

IUPAC Name: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6,9-tetraene
Synonyms: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6,9-tetraene
Molecular Weight (g/mol): 144.17
Molecular Formula: C10H8O
InChi Key: JWCGDNHAPBZVHD-UHFFFAOYNA-N
SMILES: O1C2C=CC1C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results