missing translation for 'onlineSavingsMsg'
Learn More

1,4-Epoxy-1,4-dihydronaphthalene 98.0+%, TCI America™

Catalog No. E05675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05675G 5 g
E05671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05675G Supplier TCI America Supplier No. E05675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 573-57-9
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00167584
InChI Key JWCGDNHAPBZVHD-UHFFFAOYSA-N
Synonym 1,4-epoxy-1,4-dihydronaphthalene, 1,4-dihydro-1,4-epoxynaphthalene, 1,4-epoxynaphthalene, 1,4-dihydro, 1,4-dihydronaphthalene-1,4-endo-oxide, 11-oxatricyclo 6.2.1.0 2 ,? undeca-2,4,6,9-tetraene, 11-oxatricyclo 6.2.1.0 2 ,? undeca-2 7 ,3,5,9-tetraene, 1,4-dihydro-1,4-expoxynaphthalene, 1,4-expoxynaphthalene, 1,4-dihydro, acmc-1ap1k, 1,4-dihydronaphthalene-1,4-oxide
PubChem CID 97139
SMILES C1=CC=C2C3C=CC(C2=C1)O3

Specifications

Melting Point 57°C
Color White
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Epoxy-1,4-dihydronaphthalene