accessibility menu, dialog, popup

UserName

CAS RN 5725-91-7

IUPAC Name: 7-ethoxy-3H-phenoxazin-3-one
Synonyms: ethoxyresorufin resorufin ethyl ether 7-ethoxyresorufin 7-ethoxyphenoxazone
Molecular Weight (g/mol): 241.25
Molecular Formula: C14H11NO3
InChi Key: CRCWUBLTFGOMDD-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results