missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxyresorufin, Thermo Scientific Chemicals

Catalog No. AAJ62987LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62987LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62987LB0 Supplier Thermo Scientific Chemicals Supplier No. J62987LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Fluorimetric substrate for detection of cytochrome P450 linked enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5725-91-7
Molecular Formula C14H11NO3
Molecular Weight (g/mol) 241.246
MDL Number MFCD00037661
InChI Key CRCWUBLTFGOMDD-UHFFFAOYSA-N
Synonym 7-ethoxyresorufin, ethoxyresorufin, resorufin ethyl ether, 7-ethoxy-3h-phenoxazin-3-one, 7-ethoxyphenoxazone, 3h-phenoxazin-3-one, 7-ethoxy, o7-ethylresorufin, 3h-phenoxazin-3-one,7-ethoxy, 7-ethoxy resorufin, 7-ethoxy-3-phenoxazinone
PubChem CID 3294
ChEBI CHEBI:34480
IUPAC Name 7-ethoxyphenoxazin-3-one
SMILES CCOC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2

Specifications

Melting Point 237°C to 240°C
Color Brown
Quantity 5 mg
Beilstein 225973
Formula Weight 241.24
Physical Form Powder
Chemical Name or Material 7-Ethoxyresorufin
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only