accessibility menu, dialog, popup

UserName

CAS RN 56897-54-2

IUPAC Name: dipotassium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: dipotassium eosin YS(2-)
Molecular Weight (g/mol): 724.08
Molecular Formula: C20H6Br4K2O5
InChi Key: GZAAPEKTGHKWRZ-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results