missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromofluorescein Potassium Salt 85.0+%, TCI America™

Catalog No. T003625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T003625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T003625G Supplier TCI America Supplier No. T003625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56897-54-2
Molecular Formula C20H6Br4K2O5
Molecular Weight (g/mol) 724.076
MDL Number MFCD00070510
InChI Key GZAAPEKTGHKWRZ-UHFFFAOYSA-L
PubChem CID 12598436
IUPAC Name dipotassium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].[K+].[K+]

Specifications

Quantity 25 g
Formula Weight 724.08
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromofluorescein Potassium Salt