CAS RN 56891-59-9
CAS RN 56891-59-9
IUPAC Name:
N-(4-amino-2-methylphenyl)acetamide
Synonyms:
N-(4-amino-2-methylphenyl)acetamide
Molecular Weight (g/mol):
164.21
Molecular Formula:
C9H12N2O
InChi Key:
GWFPMSIIVJMYRZ-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C(N)C=C1C
1
–
2
of
2
results
2
N1-(4-Amino-2-methylphenyl)acetamide, 90%, Thermo Scientific™
CAS: 56891-59-9 Molecular Formula: C9H12N2O Molecular Weight (g/mol): 164.208 MDL Number: MFCD00276633 InChI Key: GWFPMSIIVJMYRZ-UHFFFAOYSA-N Synonym: n-4-amino-2-methylphenyl acetamide,n1-4-amino-2-methylphenyl acetamide,4'-amino-2'-methylacetanilide,n-4-amino-2-methyl-phenyl-acetamide,4-acetamido-m-toluidine,4-acetylamino-m-toluidine,4-acetamido-3-methylaniline,4-acetamino-3-methylaniline,acetamide, n-4-amino-2-methylphenyl PubChem CID: 314338 IUPAC Name: N-(4-amino-2-methylphenyl)acetamide SMILES: CC1=C(C=CC(=C1)N)NC(=O)C