missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Amino-2-methylphenyl)acetamide, 90%, Thermo Scientific™

Catalog No. BTB06485DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB06485DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB06485DA Supplier Thermo Scientific Chemicals Supplier No. BTB06485DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56891-59-9
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
MDL Number MFCD00276633
InChI Key GWFPMSIIVJMYRZ-UHFFFAOYSA-N
Synonym n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl
PubChem CID 314338
IUPAC Name N-(4-amino-2-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)N)NC(=O)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.21
Percent Purity 90%
Chemical Name or Material N1-(4-amino-2-methylphenyl)acetamide
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.