accessibility menu, dialog, popup

UserName

CAS RN 56765-79-8

IUPAC Name: 4-aminobenzene-1,2-dicarbonitrile
Synonyms: 4-aminobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 143.15
Molecular Formula: C8H5N3
InChi Key: RRCAJFYQXKPXOJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C#N)C(=C1)C#N
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)