accessibility menu, dialog, popup

UserName

CAS RN 56613-80-0

IUPAC Name: (1S)-2-hydroxy-1-phenylethan-1-aminium
Synonyms: (1S)-2-hydroxy-1-phenylethanaminium
Molecular Weight (g/mol): 138.19
Molecular Formula: C8H12NO
InChi Key: IJXJGQCXFSSHNL-MRVPVSSYSA-O
SMILES: [NH3+][C@H](CO)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)