accessibility menu, dialog, popup

UserName

OHOOHO

CAS RN 5660-81-1

IUPAC Name: 2-cyclopentylpropanedioic acid
Synonyms: 2-cyclopentylpropanedioic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: DRHLPIHNSBNMFF-UHFFFAOYSA-N
SMILES: OC(=O)C(C1CCCC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results