accessibility menu, dialog, popup

UserName

CAS RN 5632-29-1

H3COONHBrCl

CAS RN 5632-29-1

IUPAC Name: N-(3-bromophenyl)-2-(4-chloro-3-methylphenoxy)acetamide
Synonyms: N-(3-bromophenyl)-2-(4-chloro-3-methylphenoxy)acetamide
Molecular Weight (g/mol): 354.63
Molecular Formula: C15H13BrClNO2
InChi Key: MUJGYDFUOGCMMP-UHFFFAOYSA-N
SMILES: CC1=CC(OCC(=O)NC2=CC=CC(Br)=C2)=CC=C1Cl
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results