missing translation for 'onlineSavingsMsg'
Learn More

alpha-Quaterthiophene, TCI America™

Catalog No. Q0078100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0078100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0078100MG Supplier TCI America Supplier No. Q0078100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5632-29-1
Molecular Formula C15H13BrClNO2
Molecular Weight (g/mol) 354.63
MDL Number MFCD03094036
InChI Key MUJGYDFUOGCMMP-UHFFFAOYSA-N
PubChem CID 395493
IUPAC Name N-(3-bromophenyl)-2-(4-chloro-3-methylphenoxy)acetamide
SMILES CC1=CC(OCC(=O)NC2=CC=CC(Br)=C2)=CC=C1Cl

Specifications

Melting Point 216°C
Color Yellow
Quantity 100 mg
Formula Weight 330.50
Physical Form Crystalline Powder
Chemical Name or Material alpha-Quaterthiophene