accessibility menu, dialog, popup

UserName

H3CO

CAS RN 5614-37-9

IUPAC Name: methoxycyclopentane
Synonyms: cyclopentyl methyl ether
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: SKTCDJAMAYNROS-UHFFFAOYSA-N
SMILES: COC1CCCC1
Boiling Point 
  • (5)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (9)
  • (5)
  • (6)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (2)
  • (4)