accessibility menu, dialog, popup

UserName

CAS RN 561-20-6

IUPAC Name: 2-{9-nitro-7,8-dioxo-15-oxa-1,11-diazahexacyclo[16.3.1.0⁴,¹².0⁴,²¹.0⁵,¹⁰.0¹³,¹⁹]docosa-5,9,17-trien-14-yl}acetic acid
Synonyms: cacotheline
Molecular Weight (g/mol): 427.41
Molecular Formula: C21H21N3O7
InChi Key: IVEMPCACOMNRGI-UHFFFAOYNA-N
SMILES: OC(=O)CC1OCC=C2CN3CCC45C3CC2C1C4NC1=C(C(=O)C(=O)C=C51)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results