missing translation for 'onlineSavingsMsg'
Learn More

Cacotheline Monohydrate 98.0+%, TCI America™

Catalog No. C00015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00015G 5 g
C00011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00015G Supplier TCI America Supplier No. C00015G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 561-20-6
Molecular Formula C21H21N3O7
Molecular Weight (g/mol) 427.41
MDL Number MFCD00077211
InChI Key IVEMPCACOMNRGI-UHFFFAOYNA-N
PubChem CID 101667989
IUPAC Name 2-{9-nitro-7,8-dioxo-15-oxa-1,11-diazahexacyclo[16.3.1.0⁴,¹².0⁴,²¹.0⁵,¹⁰.0¹³,¹⁹]docosa-5,9,17-trien-14-yl}acetic acid
SMILES OC(=O)CC1OCC=C2CN3CCC45C3CC2C1C4NC1=C(C(=O)C(=O)C=C51)[N+]([O-])=O

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 490.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cacotheline Monohydrate