accessibility menu, dialog, popup

UserName

CAS RN 55788-44-8

IUPAC Name: 3-bromopropane-1-sulfonic acid sodium
Synonyms: 3-bromopropane-1-sulfonic acid sodium
Molecular Weight (g/mol): 226.04
Molecular Formula: C3H7BrNaO3S
InChi Key: ONPNLMMBTJZJRQ-UHFFFAOYSA-N
SMILES: [Na].OS(=O)(=O)CCCBr
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)