missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Bromopropanesulfonate 98.0+%, TCI America™

Catalog No. B170725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B170725G 25 g
B1707100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B170725G Supplier TCI America Supplier No. B170725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55788-44-8
Molecular Formula C3H7BrNaO3S
Molecular Weight (g/mol) 226.04
MDL Number MFCD00024999
InChI Key ONPNLMMBTJZJRQ-UHFFFAOYSA-N
Synonym sodium 3-bromopropanesulfonate, sodium 3-bromopropane-1-sulfonate, 3-bromopropanesulfonic acid sodium salt, acmc-209lpd, sodium 3-bromo-1-propanesulfonate, sodium 3-bromo-propane-1-sulphonate, 3-bromopropane sulfonic acid sodium salt, 3-bromo-1-propanesulfonic acid sodium salt, 1-propanesulfonic acid, 3-bromo-, sodium salt
PubChem CID 23679056
IUPAC Name 3-bromopropane-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)CCCBr

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 225.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Bromopropanesulfonate