accessibility menu, dialog, popup

UserName

CAS RN 55750-06-6

IUPAC Name: 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea; propanoic acid
Synonyms: imidocarb; propanoic acid
Molecular Weight (g/mol): 422.49
Molecular Formula: C22H26N6O3
InChi Key: IRPJGGHUXQZOQW-UHFFFAOYSA-N
SMILES: CCC(O)=O.O=C(NC1=CC=CC(=C1)C1=NCCN1)NC1=CC=CC(=C1)C1=NCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results