accessibility menu, dialog, popup

UserName

CAS RN 55663-99-5

IUPAC Name: zinc(2+) bis(N-[4-(diazyn-1-ium-1-yl)-2,5-dimethoxyphenyl]benzenecarboximidate) dihydrochloride dichloride
Synonyms: zinc(2+) bis(N-[4-(diazyn-1-ium-1-yl)-2,5-dimethoxyphenyl]benzenecarboximidate) dihydrochloride dichloride
Molecular Weight (g/mol): 775.77
Molecular Formula: C30H28Cl4N6O6Zn
InChi Key: LLARVKAOQLUCQI-UHFFFAOYSA-L
SMILES: Cl.Cl.[Cl-].[Cl-].[Zn++].COC1=CC([N+]#N)=C(OC)C=C1N=C([O-])C1=CC=CC=C1.COC1=CC([N+]#N)=C(OC)C=C1N=C([O-])C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results