accessibility menu, dialog, popup

UserName

ONH2OHHO

CAS RN 556-03-6

IUPAC Name: 2-amino-3-(4-hydroxyphenyl)propanoic acid
Synonyms: tyrosin|tirosina
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: OUYCCCASQSFEME-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)