missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tyrosine 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 181.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Tyrosine |
Safety and Handling
| EINECSNumber | (9)-1596 |
| TSCA | Yes |