accessibility menu, dialog, popup

UserName

ONOO

CAS RN 555-16-8

IUPAC Name: 4-nitrobenzaldehyde
Synonyms: 4-nitrobenzaldehyde
Molecular Weight (g/mol): 151.12
Molecular Formula: C7H5NO3
InChi Key: BXRFQSNOROATLV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=O)C=C1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)