missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. N0559500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0559500G 500 g
N055925G 25 g
N0559100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0559500G Supplier TCI America Supplier No. N0559500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 555-16-8
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007346
InChI Key BXRFQSNOROATLV-UHFFFAOYSA-N
Synonym p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
PubChem CID 541
ChEBI CHEBI:66926
IUPAC Name 4-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 105°C
Quantity 500 g
Formula Weight 151.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzaldehyde
EINECSNumber (3)-1181
RTECSNumber CU7350000
TSCA Yes