accessibility menu, dialog, popup

UserName

CAS RN 55406-53-6

IUPAC Name: 3-iodoprop-2-yn-1-yl N-butylcarbamate
Synonyms: iodopropynyl butylcarbamate
Molecular Weight (g/mol): 281.09
Molecular Formula: C8H12INO2
InChi Key: WYVVKGNFXHOCQV-UHFFFAOYSA-N
SMILES: CCCCNC(=O)OCC#CI
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)