missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-propynyl N-butylcarbamate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111016544
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016544 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016544 Supplier Merck Emd Millipore Supplier No. PHR1354500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Iodo-2-propynyl N-butylcarbamate is widely used as an antimicrobial and antifungal agent. It can be encapsulated in polystrene/polycaprolactone blend in the form microspheres. The microspheres were tested for invitro antifouling activity. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 55406-53-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material