missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-propynyl N-butylcarbamate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019268
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019268 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019268 Supplier Merck Emd Millipore Supplier No. 90865100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 55406-53-6
Physical Form Neat
Linear Formula CH3(CH2)3 NHCO2 CH2 C///Cl
MDL Number MFCD00072438
Melting Point 64°C to 68°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 281.09
Percent Purity ≥98.5% (GC)
Grade Analytical Standard