missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-2-propynyl N-Butylcarbamate 97.0+%, TCI America™

Catalog No. I066625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I066625G 25 g
I0666100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I066625G Supplier TCI America Supplier No. I066625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55406-53-6
Molecular Formula C8H12INO2
Molecular Weight (g/mol) 281.093
MDL Number MFCD00072438
InChI Key WYVVKGNFXHOCQV-UHFFFAOYSA-N
Synonym 3-iodoprop-2-yn-1-yl butylcarbamate, iodopropynyl butylcarbamate, iodocarb, ipbc, woodlife, 3-iodo-2-propynyl butylcarbamate, 1-iodoprop-1-yn-3-yl n-n-butylcarbamate, troysan kk-108a, carbamic acid, butyl-, 3-iodo-2-propynyl ester, troysan polyphase anti-mildew
PubChem CID 62097
ChEBI CHEBI:83279
IUPAC Name 3-iodoprop-2-ynyl N-butylcarbamate
SMILES CCCCNC(=O)OCC#CI

Specifications

Melting Point 69°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 281.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-2-propynyl N-Butylcarbamate
EINECSNumber (2)-3456
RTECSNumber EZ0950000
TSCA Yes