accessibility menu, dialog, popup

UserName

CH3OOH3C

CAS RN 554-12-1

IUPAC Name: methyl propanoate
Synonyms: methyl propionate
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: RJUFJBKOKNCXHH-UHFFFAOYSA-N
SMILES: CCC(=O)OC
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)