missing translation for 'onlineSavingsMsg'
Learn More

Methyl propionate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020246
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020246 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020246 Supplier Merck Emd Millipore Supplier No. 819881ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 554-12-1
Ignition Point 876°F
Vapor Pressure 85.3 mmHg (25°C)
Vapor Density 3 (vs. air)
Density 0.915 g/mL (at 25°C)
Flash Point -2°C
Physical Form Neat
Linear Formula CH3 CH2 COOCH3
MDL Number MFCD00009306
Melting Point -88°C
Boiling Point 79°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 88.11
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less