missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propionate 99.5+%, TCI America™

Catalog No. S03015ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03015ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03015ML Supplier TCI America Supplier No. S03015ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 554-12-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009306
InChI Key RJUFJBKOKNCXHH-UHFFFAOYSA-N
Synonym methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
IUPAC Name methyl propanoate
SMILES CCC(=O)OC

Specifications

Melting Point -88°C
Color Colorless
Boiling Point 80°C
Quantity 5 mL
UN Number 1248
Formula Weight 88.11
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Propionate [Standard Material for GC]
EINECSNumber (2)-0774
RTECSNumber UF5970000
TSCA Yes