missing translation for 'onlineSavingsMsg'
Learn More

Methyl propionate, 99%

Catalog No. AAA14944AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14944AP 500 mL
AAA14944AE 100 mL
AAA149440F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14944AP Supplier Thermo Scientific Chemicals Supplier No. A14944AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 554-12-1 | C4H8O2 | 88.106 g/mol

It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 554-12-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009306
InChI Key RJUFJBKOKNCXHH-UHFFFAOYSA-N
Synonym methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
IUPAC Name methyl propanoate
SMILES CCC(=O)OC

Specifications

Melting Point -88°C
Density 0.915
Boiling Point 78°C to 80°C
Flash Point −2°C (28°F)
Assay Percent Range 99%
Refractive Index 1.377
Quantity 500 mL
UN Number UN1248
Beilstein 1737628
Merck Index 14,6112
Solubility Information Soluble in water (5g/L).
Formula Weight 88.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl propionate
Show More Show Less
Hazard Category H225-H315-H319-H332
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPIONATE
EINECSNumber 209-060-4
RTECSNumber UF5970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only