accessibility menu, dialog, popup

UserName

CAS RN 55216-11-0

CH3O(R)O(R)O(R)(R)OH3CO(R)O(R)(R)OH3CO(R)O(R)(R)OCH3O(R)O(R)(R)OCH3O(R)O(R)(R)OH3CO(R)O(R)(R)OH3CO(R)O(R)(S)OCH3(R)OCH3(R)OCH3(S)OCH3(R)OCH3(S)OCH3(R)OCH3(S)OH3C(R)OH3C(S)OH3C(R)OCH3(S)OH3C(R)OH3C(S)OH3C

CAS RN 55216-11-0

IUPAC Name: (1R,3R,5R,6R,8R,10R,11R,13R,15R,16R,18R,20R,21R,23R,25R,26R,28R,30R,31R,33R,35R,36S,37R,38S,39R,40S,41R,42S,43R,44S,45R,46S,47R,48S,49R)-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane
Synonyms: (1R,3R,5R,6R,8R,10R,11R,13R,15R,16R,18R,20R,21R,23R,25R,26R,28R,30R,31R,33R,35R,36S,37R,38S,39R,40S,41R,42S,43R,44S,45R,46S,47R,48S,49R)-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane
Molecular Weight (g/mol): 1429.55
Molecular Formula: C63H112O35
InChi Key: DSDAICPXUXPBCC-MWDJDSKUSA-N
SMILES: COC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC)O[C@H](O[C@@H]4[C@@H](COC)O[C@H](O[C@@H]5[C@@H](COC)O[C@H](O[C@@H]6[C@@H](COC)O[C@H](O[C@@H]7[C@@H](COC)O[C@H](O[C@@H]8[C@@H](COC)O[C@H](O[C@H]1[C@H](OC)[C@H]2OC)[C@H](OC)[C@H]8OC)[C@H](OC)[C@H]7OC)[C@H](OC)[C@H]6OC)[C@H](OC)[C@H]5OC)[C@H](OC)[C@H]4OC)[C@H](OC)[C@H]3OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results