missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-beta-cyclodextrin 98.0+%, TCI America™

Catalog No. T10941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10941G Supplier TCI America Supplier No. T10941G
Only null left

Chemical Identifiers

CAS 55216-11-0
Molecular Formula C63H112O35
Molecular Weight (g/mol) 1429.55
MDL Number MFCD00010728
InChI Key DSDAICPXUXPBCC-MWDJDSKUSA-N
PubChem CID 133126616
IUPAC Name (1R,3R,5R,6R,8R,10R,11R,13R,15R,16R,18R,20R,21R,23R,25R,26R,28R,30R,31R,33R,35R,36S,37R,38S,39R,40S,41R,42S,43R,44S,45R,46S,47R,48S,49R)-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane
SMILES COC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC)O[C@H](O[C@@H]4[C@@H](COC)O[C@H](O[C@@H]5[C@@H](COC)O[C@H](O[C@@H]6[C@@H](COC)O[C@H](O[C@@H]7[C@@H](COC)O[C@H](O[C@@H]8[C@@H](COC)O[C@H](O[C@H]1[C@H](OC)[C@H]2OC)[C@H](OC)[C@H]8OC)[C@H](OC)[C@H]7OC)[C@H](OC)[C@H]6OC)[C@H](OC)[C@H]5OC)[C@H](OC)[C@H]4OC)[C@H](OC)[C@H]3OC

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 1429.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl-beta-cyclodextrin